Novus Biologicals
Manufacturer Code:NBP2844800.1ML
Catalog # NB032408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human ATP5D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase F1 subunit delta; ATP synthase subunit delta mitochondrial ATP synthase H+ transporting mitochondrial F1 complex delta subunit F-ATPase delta subunit mitochondrial ATP synthase complex delta-subunit precusor mitochondrial ATP synthase delta subunit; ATP synthase subunit delta, mitochondrial; F-ATPase delta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase, delta subunit
Gene Aliases: ATP5D; ATP5F1D
UniProt ID: (Human) P30049
Entrez Gene ID: (Human) 513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.