Novus Biologicals
Manufacturer Code:NB10093582SS
Catalog # NB017930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Reacts with residues 33-43 [CGVYLLPRRGPR] of HCV core enf and part of E2/NS1 of HCV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: asparagine synthetase domain containing 1 asparagine synthetase domain-containing protein 1 HCV NS3-transactivated protein 1 NBLA00058 NS3TP1FLJ20752; Asparagine synthetase domain-containing protein 1; HCV NS3-transactivated protein 1
Gene Aliases: ASNSD1; NBLA00058; NS3TP1
UniProt ID: (Human) Q9NWL6
Entrez Gene ID: (Human) 54529
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.