Novus Biologicals
Manufacturer Code:NBP270205V
Catalog # NBP270205V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G5 |
Immunogen | Full length recombinant protein of human ARG2 (NP_001163) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: arginase 2 arginase type II arginase-2 mitochondrial EC 3.5.3 EC 3.5.3.1 kidney arginase Kidney-type arginase L-arginine amidinohydrolase L-arginine ureahydrolase nonhepatic arginase Non-hepatic arginase Type II arginase; Arginase II; arginase, type II; Arginase-2, mitochondrial; kidney arginase; Kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; Non-hepatic arginase; nonhepatic arginase; Type II arginase
Gene Aliases: ARG2
UniProt ID: (Human) P78540
Entrez Gene ID: (Human) 384
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.