Novus Biologicals
Manufacturer Code:NBP154389
Catalog # NBP154389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ARG2(arginase type II) The peptide sequence was selected from the C terminal of ARG2. Peptide sequence SALDLVEVNPQLATSEEEAKTTANLAVDVIASSFGQTREGGHIVYDQLPT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arginase 2 arginase type II arginase-2 mitochondrial EC 3.5.3 EC 3.5.3.1 kidney arginase Kidney-type arginase L-arginine amidinohydrolase L-arginine ureahydrolase nonhepatic arginase Non-hepatic arginase Type II arginase; Arginase II; arginase, type II; Arginase-2, mitochondrial; kidney arginase; Kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; Non-hepatic arginase; nonhepatic arginase; Type II arginase
Gene Aliases: ARG2
UniProt ID: (Human) P78540
Entrez Gene ID: (Human) 384
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.