Invitrogen
Manufacturer Code:PA513797
Catalog # PIPA513797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 148-178 amino acids from the central region of human PRKAG3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-AMP-activated protein kinase gamma-3 subunit AMPK gamma-3 chain AMPK gamma3 AMPKG3; 5'-AMP-activated protein kinase subunit gamma-3; 5'-AMP-activated protein kinase, gamma-3 subunit; AMP-activated protein kinase gamma subunit; AMPK gamma-3 chain; AMPK gamma3; AMPK subunit gamma-3; protein kinase, AMP-activated, gamma 3 non-catalytic subunit
Gene Aliases: Amkg3; AMPKG3; AMPKg3L; AMPKg3S; PRKAG3
UniProt ID: (Human) Q9UGI9, (Mouse) Q8BGM7
Entrez Gene ID: (Human) 53632, (Mouse) 241113
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.