Invitrogen
Manufacturer Code:MA535486
Catalog # PIMA535486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1061 |
Immunogen | A synthesized peptide derived from human AMPK beta 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 1300015D22Rik 5&prime-AMP-activated protein kinase 40 kDa subunit 5&prime-AMP-activated protein kinase subunit beta-1 AAKB1 AMPK AMPK subunit beta-1 AMPK-beta1 AU021155 E430008F22 HAMPKb kinase AMPK-beta1 kinase pAMPK pAMPK Prkab1; 5'-AMP-activated protein kinase 40 kDa subunit; 5'-AMP-activated protein kinase subunit beta-1; 5'-AMP-activated protein kinase, beta subunit; 5-AMP-activated protein kinase beta subunit; AMPK beta-1 chain; AMPK subunit beta-1; AMPKb
Gene Aliases: 1300015D22Rik; AU021155; E430008F22; Prkab1
UniProt ID: (Mouse) Q9R078, (Rat) P80386
Entrez Gene ID: (Mouse) 19079, (Rat) 83803
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.