Invitrogen
Manufacturer Code:PA514108
Catalog # PIPA514108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 474-502 amino acids from human AMPK alpha(PRKAA1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK alpha 1; AMPK subunit alpha-1; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; Protein kinase AMP-activated alpha 1 catalytic subunit AMPK alpha-1 chain; protein kinase, AMP-activated, alpha 1 catalytic subunit; Tau-protein kinase PRKAA1
Gene Aliases: AMPK; AMPK1; AMPKa1; PRKAA1
UniProt ID: (Human) Q13131
Entrez Gene ID: (Human) 5562
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.