Novus Biologicals
Manufacturer Code:NBP2444410.2MG
Catalog # NBP24444102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AMPD3/901 |
Immunogen | Recombinant full-length human AMDP3 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase (isoform E); adenosine monophosphate deaminase (isoform E) adenosine monophosphate deaminase 3 AMP aminohydrolase AMP deaminase 3 AMP deaminase isoform E EC 3.5.4.6 Erythrocyte AMP deaminase erythrocyte type AMP deaminase erythrocyte-specific AMP deaminase myoadenylate deaminase; AMP aminohydrolase; AMP deaminase 3; AMP deaminase isoform E; Erythrocyte AMP deaminase; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; myoadenylate deaminase
Gene Aliases: AMPD3
UniProt ID: (Human) Q01432
Entrez Gene ID: (Human) 272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.