Abnova
Manufacturer Code:MAB13288
Catalog # 89-933-565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AMPD3/901 |
Immunogen | Recombinant protein corresponding to full length human AMPD3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full length recombinant human AMPD3.
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase (isoform E); AMP aminohydrolase; AMP deaminase 3; AMP deaminase isoform E; Erythrocyte AMP deaminase; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; myoadenylate deaminase
Gene Aliases: AMPD3
UniProt ID: (Human) Q01432
Entrez Gene ID: (Human) 272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.