Invitrogen
Manufacturer Code:PA515032
Catalog # PIPA515032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 325-356 amino acids from the central region of human AMPD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase (isoform E); AMP aminohydrolase; AMP deaminase 3; AMP deaminase H-type; AMP deaminase isoform E; Erythrocyte AMP deaminase; Erythrocyte AMP deaminase AMP deaminase isoform E AMP deaminase 3 AMPD3; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; Heart-type AMPD; myoadenylate deaminase
Gene Aliases: AMPD3
UniProt ID: (Human) Q01432, (Mouse) O08739
Entrez Gene ID: (Human) 272, (Mouse) 11717
Molecular Function: deaminase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.