Invitrogen
Manufacturer Code:PA5117108
Catalog # PIPA5117108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AMPD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 1 (isoform M); adenosine monophosphate deaminase 1 adenosine monophosphate deaminase 1 (isoform M) adenosine monophosphate deaminase-1 (muscle) AI553520 AMP deaminase 1 AMP deaminase isoform M AMPD Ampd01 Ampd1 Ampd-1 MAD MADA MMDD myoadenylate deaminase RATAMPD01 RP5-1000E10.1 skeletal muscle AMPD; adenosine monophosphate deaminase-1 (muscle); AMP deaminase 1; AMP deaminase isoform M; AMPD; Myoadenylate deaminase; skeletal muscle AMPD
Gene Aliases: AI553520; Ampd-1; Ampd01; AMPD1; MAD; MADA; MMDD; RATAMPD01
UniProt ID: (Human) P23109, (Rat) P10759, (Mouse) Q3V1D3
Entrez Gene ID: (Human) 270, (Rat) 25028, (Mouse) 229665
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.