OriGene
Manufacturer Code:CF801141
Catalog # 50-166-5465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1060-1620 of human ALK produced in SF9 Cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ALK tyrosine kinase receptor; Anaplastic lymphoma kinase; CD246; CD246 antigen; CD246 NBLST3; mutant anaplastic lymphoma kinase
Gene Aliases: ALK; CD246; NBLST3
UniProt ID: (Human) Q9UM73
Entrez Gene ID: (Human) 238
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.