Invitrogen
Manufacturer Code:PA575265
Catalog # PIPA575265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminual of Human AK6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate kinase isoenzyme 6; Adenylate kinase isoenzyme 6 AK6 Adrenal gland protein AD-004 Coilin-interacting nuclear ATPase protein hCINAP Dual activity adenylate kinase/ATPase AK/ATPase CINAP AD-004 CGI-137; adenylate kinase isoenzyme 6 {ECO:0000255|HAMAP-Rule:MF_03173}; Adrenal gland protein AD-004; AK/ATPase; AK/ATPase {ECO:0000255|HAMAP-Rule:MF_03173}; AK6; AK6 {ECO:0000255|HAMAP-Rule:MF_03173}; ATP-AMP transphosphorylase 6; coilin interacting nuclear ATPase protein; coilin interacting protein; Coilin-interacting nuclear ATPase protein; coilin-interacting nuclear ATPase protein {ECO:0000255|HAMAP-Rule:MF_03173}; Dual activity adenylate kinase/ATPase; dual activity adenylate kinase/ATPase {ECO:0000255|HAMAP-Rule:MF_03173}; hCINAP
Gene Aliases: 2810046E22Rik; 4921516M08Rik; AD-004; AK/ATPase; AK6; CGI-137; CINAP; CIP; hCINAP
UniProt ID: (Human) Q9Y3D8, (Rat) Q5EB68, (Mouse) Q8VCP8
Entrez Gene ID: (Human) 102157402, (Rat) 102238592, (Mouse) 102216272
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.