Novus Biologicals
Manufacturer Code:NBP262219R
Catalog # NB002719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AGPS-03 |
Immunogen | recombinant human AGPS (amino acids 158-384) |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ADAP-S ADAS ADHAPSAlkylglycerone-phosphate synthase ADPS Aging-associated gene 5 protein aging-associated protein 5 ALDHPSY Alkyl-DHAP synthase alkyldihydroxyacetonephosphate synthase peroxisomal alkylglycerone phosphate synthase DKFZp762O2215 EC 2.5.1.26 FLJ99755; Aging-associated gene 5 protein; aging-associated protein 5; Alkyl-DHAP synthase; Alkyldihydroxyacetonephosphate synthase, peroxisomal; Alkylglycerone-phosphate synthase
Gene Aliases: AAG5; ADAP-S; ADAS; ADHAPS; ADPS; AGPS; ALDHPSY; RCDP3
UniProt ID: (Human) O00116
Entrez Gene ID: (Human) 8540
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.