Novus Biologicals
Manufacturer Code:NBP2825830.1ML
Catalog # NB030512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human ADPRHL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Protein ADP-ribosylarginine] hydrolase-like protein 1; [Protein ADP-ribosylarginine] hydrolase-like protein 1 ADP-ribosyl-hydrolase ADP-ribosylhydrolase like 1 ARH2ADP-ribosylhydrolase 2; ADP-ribosyl-hydrolase; ADP-ribosylarginine hydrolase like 1; ADP-ribosylhydrolase 2
Gene Aliases: ADPRHL1; ARH2
UniProt ID: (Human) Q8NDY3
Entrez Gene ID: (Human) 113622
Molecular Function: G-protein modulator enzyme modulator hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.