Invitrogen
Manufacturer Code:PA531416
Catalog # PIPA531416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 105 and 328 of Human ADHFE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH8 HOT Fe-containing alcohol dehydrogenase 1 alcohol dehydrogenase 8 alcohol dehydrogenase iron-containing protein 1 hydroxyacid-oxoacid transhydrogenase mitochondrial; ADHFe1; alcohol dehydrogenase 8; Alcohol dehydrogenase iron-containing protein 1; alcohol dehydrogenase, iron containing, 1; Fe-containing alcohol dehydrogenase; Fe-containing alcohol dehydrogenase 1; HOT; Hydroxyacid-oxoacid transhydrogenase, mitochondrial
Gene Aliases: ADH8; ADHFE1; HMFT2263; HOT
UniProt ID: (Human) Q8IWW8
Entrez Gene ID: (Human) 137872
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.