Abnova
Manufacturer Code:H00137872K
Catalog # 89-204-728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ADHFE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ADHFE1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ADH8 FLJ32430 HMFT2263 HOT MGC48605; ADHFe1; alcohol dehydrogenase 8; Alcohol dehydrogenase iron-containing protein 1; alcohol dehydrogenase, iron containing, 1; Fe-containing alcohol dehydrogenase; Fe-containing alcohol dehydrogenase 1; HOT; Hydroxyacid-oxoacid transhydrogenase, mitochondrial
Gene Aliases: ADH8; ADHFE1; HMFT2263; HOT
UniProt ID: (Human) Q8IWW8
Entrez Gene ID: (Human) 137872
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.