Invitrogen
Manufacturer Code:PA5102833
Catalog # PIPA5102833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ADH6 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810009C16Rik ADH-5 Adh5a Adh6 Adh6a alcohol dehydrogenase 6 alcohol dehydrogenase 6 (class V) alcohol dehydrogenase 6A (class V) aldehyde reductase class V alcohol dehydrogenase; Alcohol dehydrogenase 6; alcohol dehydrogenase 6A (class V); aldehyde reductase; class V alcohol dehydrogenase
Gene Aliases: 1810009C16Rik; ADH-5; Adh5a; ADH6
UniProt ID: (Human) P28332, (Rat) Q5XI95
Entrez Gene ID: (Human) 130, (Rat) 310903, (Mouse) 69117, (Rat) 295498
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.