Novus Biologicals
Manufacturer Code:NBP185508
Catalog # NBP185508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RQSADFMPRWQMLRVLEEELGRDWQAKVASLEEVPFAAASIGQVHQGLLRDGTEVAVKIQY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aarF domain containing kinase 4; aarF domain containing kinase 4 COQ8 EC 2.7.11 EC 2.7.11.- EC 2.7.11.1 EC 2.7.11.25 FLJ12229; AarF domain-containing protein kinase 4; Atypical kinase COQ8B, mitochondrial; Coenzyme Q protein 8B
Gene Aliases: ADCK4; COQ8B; NPHS9
UniProt ID: (Human) Q96D53
Entrez Gene ID: (Human) 79934
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.