Proteintech
Manufacturer Code:249341AP150UL
Catalog # 50-173-4626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADAMTS12 Fusion Protein Ag20712 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 12; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 12; a disintegrin-like and metalloprotease with thrombospondin type 1 motif, 12; ADAM metallopeptidase with thrombospondin type 1 motif, 12; ADAM TS 12 ADAM TS12 ADAMTS 12 ADAMTS12 PRO4389; ADAM-TS 12; ADAM-TS12; ADAMTS-12; metalloprotease disintegrin 12
Gene Aliases: ADAMTS-12; ADAMTS12; AI605170; PRO4389; UNQ1918/PRO4389
UniProt ID: (Human) A2RRN9, (Mouse) Q811B3
Entrez Gene ID: (Human) 81792, (Mouse) 239337
Molecular Function: enzyme modulator extracellular matrix glycoprotein extracellular matrix protein hydrolase metalloprotease protease protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.