Invitrogen
Manufacturer Code:PA5113492
Catalog # PIPA5113492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Lysophosphatidic acid phosphatase type 6 protein (35-324AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3.1.3.2 5730559A09Rik Acid phosphatase 6 lysophosphatidic acid phosphatase like 1 Acid phosphatase-like protein 1 Acp6 ACPL1 AU022842 Lpap Lysophosphatidic acid phosphatase type 6 mPACPL1 PACPL1 UNQ205/PRO231; Acid phosphatase 6, lysophosphatidic; acid phosphatase like 1; Acid phosphatase-like protein 1; Lysophosphatidic acid phosphatase type 6; PACPL1
Gene Aliases: 5730559A09Rik; ACP6; ACPL1; AU022842; LPAP; mPACPL1; PACPL1; UNQ205/PRO231
UniProt ID: (Human) Q9NPH0, (Mouse) Q3TNE2
Entrez Gene ID: (Human) 51205, (Rat) 295305, (Mouse) 66659
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.