Invitrogen
Manufacturer Code:PA529956
Catalog # PIPA529956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 64 and 304 of Human ACAA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-CoA thiolase A, peroxisomal; 3-ketoacyl-CoA thiolase, peroxisomal; ACAA PTHIO THIO 3-ketoacyl-CoA thiolase peroxisomal acetyl-Coenzyme A acyltransferase 1 beta-ketothiolase peroxisomal 3-oxoacyl-CoA thiolase peroxisomal 3-oxoacyl-Coenzyme A thiolase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 1A; acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A 1 peroxisomal; Acetyl-CoA acyltransferase 3-oxo acyl-CoA thiolase A peroxisomal; Acetyl-CoA acyltransferase A; acetyl-CoA acyltransferase, 3-oxo acyl-CoA thiolase A; Acetyl-CoA C-myristoyltransferase; acetyl-Coenzyme A acyltransferase 1; acetyl-Coenzyme A acyltransferase 1 (peroxisomal 3-oxoacyl-Coenzyme A thiolase); acetyl-Coenzyme A acyltransferase 1A; Beta-ketothiolase; Beta-ketothiolase A; peroxisomal 3-ketoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase; Peroxisomal 3-oxoacyl-CoA thiolase A; peroxisomal 3-oxoacyl-Coenzyme A thiolase; testicular tissue protein Li 197; Thiolase A
Gene Aliases: ACAA; ACAA1; Acaa1a; D9Ertd25e; Pktaa; PTHIO; PTL; THIO
UniProt ID: (Human) P09110, (Mouse) Q921H8, (Rat) P21775
Entrez Gene ID: (Human) 30, (Mouse) 113868, (Rat) 24157
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.