Novus Biologicals
Manufacturer Code:NBP271540AF350
Catalog # NP27140A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C8 |
Immunogen | Full length recombinant protein of human ABCC5 (NP_001018881) produced in E.coli. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ATP-binding cassette sub-family C member 5; ATP-binding cassette sub-family C member 5 ATP-binding cassette sub-family C (CFTR/MRP) member 5 canalicular multispecific organic anion transporter C EST277145 MOAT-CABC33 MRP5MOATC multidrug resistance-associated protein 5 Multi-specific organic anion transporter C pABC11 SMRPDKFZp686C1782; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; canalicular multispecific organic anion transporter C; MOAT-C; Multi-specific organic anion transporter C; Multidrug resistance-associated protein 5; pABC11; SMRP
Gene Aliases: ABC33; ABCC5; EST277145; MOAT-C; MOATC; MRP5; pABC11; SMRP
UniProt ID: (Human) O15440
Entrez Gene ID: (Human) 10057
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.