Abnova
Manufacturer Code:H00022848K
Catalog # 89-201-070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AAK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human AAK1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Adaptor-associated kinase 1; AP2-associated protein kinase 1; DKFZp686F03202 DKFZp686K16132 FLJ23712 FLJ25931 FLJ31060 FLJ42882 FLJ45252 KIAA1048 MGC138170 MGC164568 MGC164570
Gene Aliases: AAK1; KIAA1048
UniProt ID: (Human) Q2M2I8
Entrez Gene ID: (Human) 22848
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.