Invitrogen
Manufacturer Code:PA5109351
Catalog # PIPA5109351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 170-399 of human AADAC (NP_001077.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5033417E09Rik Aada AADAC AI265437 Arylacetamide deacetylase CES5A1 DAC; Arylacetamide deacetylase; arylacetamide deacetylase (esterase)
Gene Aliases: 5033417E09Rik; Aada; AADAC; AI265437; CES5A1; DAC
UniProt ID: (Human) P22760, (Mouse) Q99PG0
Entrez Gene ID: (Human) 13, (Mouse) 67758
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.