Novus Biologicals
Manufacturer Code:NBP299385G
Catalog # NB060271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human AADAC. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arylacetamide deacetylase; arylacetamide deacetylase (esterase); arylacetamide deacetylase arylacetamide deacetylase (esterase) CES5A1 DACEC 3.1.1.3 EC 3.1.1 EC 3.1.1.79
Gene Aliases: AADAC; CES5A1; DAC
UniProt ID: (Human) P22760
Entrez Gene ID: (Human) 13
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.