Abnova
Manufacturer Code:H00026061B01
Catalog # 89-007-369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHYH2 (NP_036392.2 1 a.a. ~ 578 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PHYH2 protein.
For Research Use Only
Protein Aliases: 1600020H07Rik; 2-HPCL; 2-hydroxyacyl-CoA lyase 1; 2-hydroxyphytanol-CoA lyase; 2-hydroxyphytanoyl-CoA lyase; HPCL; HPCL2; PHYH2; Phytanoyl-CoA 2-hydroxylase 2; phytanoyl-CoA hydroxylase 2
Gene Aliases: 2-HPCL; HACL1; HPCL; HPCL2; HSPC279; PHYH2
UniProt ID: (Human) Q9UJ83
Entrez Gene ID: (Human) 26061
Molecular Function: decarboxylase dehydrogenase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.